Läs mer
(S)-3-Amino-2-azepanone (>90%), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-3-Amino-2-azepanone |
| CAS | 21568-87-6 |
| Molekylformel | C6H12N2O |
| Kvantitet | 500 mg |
| Synonym | (3S)-3-Aminohexahydro-2H-azepin-2-one, (-)-3-Aminohexahydro-2-azepinone, (-)-alpha-Amino-epsilon-caprolactam, (-)-alpha-Aminocaprolactam, (3S)-Amino-epsilon-caprolactam, (S)-(-)-alpha-Amino-epsilon-caprolactam, (S)-3-Amino-2-caprolactam, (S)-3-Aminoazepan-2-one, (S)-3-Aminohexahydro-2-azepinone, (S)-Caprolactam-3-amine, (S)-alpha-Amino-epsilon-caprolactam, (S)-alpha-Aminocaprolactam, L-(-)-alpha-Amino-epsilon-caprolactam, L-alpha-Amino-epsilon-caprolactam, L-alpha-Aminocaprolactam |
| InChI formel | InChI=1S/C6H12N2O/c7-5-3-1-2-4-8-6(5)9/h5H,1-4,7H2,(H,8,9)/t5-/m0/s1 |
| LEDER | N[C@H]1CCCCNC1=O |
| IUPAC-namn | (3S)-3-aminoazepan-2-one |
| Molekylvikt (g/mol) | 128.17 |