Learn More
Alfa Aesar™ (S)-3-Benzyloxycarbonylamino-2-azepanone, 97%
Brand:  Alfa Aesar™ H63379.03
Additional Details : CAS Number : 103478-12-2 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15470039
  • 306.60 SEK / 1g
Estimated Shipment date
from Supplier 07-03-2018
Add to basket
Specifications
Chemical Identifiers
(S)-3-Benzyloxycarbonylamino-2-azepanone
262.3
(S)-Benzyl (2-oxoazepan-3-yl)carbamate
103478-12-2
MFCD08060107
Specifications
(S)-3-Benzyloxycarbonylamino-2-azepanone
Solid
262.3
White
(S)-Benzyl (2-oxoazepan-3-yl)carbamate
103478-12-2
97%
MFCD08060107
1g