Läs mer
(S)-3-Chloro-1-phenyl-1-[2-methyl-phenoxyl]propane, TRC
Produktkod. 30367232
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367232 50 mg
30367424 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367232 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37956350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-3-Chloro-1-phenyl-1-[2-methyl-phenoxyl]propane
CAS 114446-50-3
Molekylformel C16 H17 Cl O
Kvantitet 50 mg
InChI formel InChI=1S/C16H17ClO/c1-13-7-5-6-10-15(13)18-16(11-12-17)14-8-3-2-4-9-14/h2-10,16H,11-12H2,1H3/t16-/m0/s1
LEDER Cc1ccccc1O[C@@H](CCCl)c2ccccc2
IUPAC-namn 1-[(1S)-3-chloro-1-phenylpropoxy]-2-methylbenzene
Molekylvikt (g/mol) 260.76
Formel vikt 260.0968