missing translation for 'onlineSavingsMsg'
Läs mer

(S)-3-Methylheptanoic Acid-d3, TRC

Produktkod. 30465549
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30465549 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30465549 Supplier Toronto Research Chemicals Supplier No. TRCM31193750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (S)-3-Methylheptanoic Acid-d3
CAS 1329647-33-7
Molekylformel C8H13D3O2
Kvantitet 50 mg
Synonym (S)-(-)-3-Methylheptanoic Acid-d3, (S)-3-Methyl-heptanoic Acid-d3
InChI formel InChI=1S/C8H16O2/c1-3-4-5-7(2)6-8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/t7-/m0/s1/i2D3
LEDER [2H]C([2H])([2H])[C@@H](CCCC)CC(=O)O
IUPAC-namn (3S)-3-(trideuteriomethyl)heptanoic acid
Molekylvikt (g/mol) 147.23
Formel vikt 147.1339
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.