missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Methylmorpholine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7077641
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1022094-03-6
C5H11NO·ClH
137.61
MSOCQCWIEBVSLF-JEDNCBNOSA-N
s-3-methylmorpholine hydrochloride, s-3-methylmorpholine hcl, 3s-3-methylmorpholine hydrochloride, s-3-methyl-morpholine hydrochloride, s-3-methyl morpholine hydrochloride, c5h11no.clh, s-3-methylmorpholinehydrochloride, s-3-methylmorpholin hydrochloride, 3s-3-methylmorpholine-hydrogen chloride 1/1
53486469
(3S)-3-methylmorpholine;hydrochloride
CC1COCCN1.Cl
This item is not returnable. View return policy
1022094-03-6
C5H11NO·ClH
137.61
MSOCQCWIEBVSLF-JEDNCBNOSA-N
s-3-methylmorpholine hydrochloride, s-3-methylmorpholine hcl, 3s-3-methylmorpholine hydrochloride, s-3-methyl-morpholine hydrochloride, s-3-methyl morpholine hydrochloride, c5h11no.clh, s-3-methylmorpholinehydrochloride, s-3-methylmorpholin hydrochloride, 3s-3-methylmorpholine-hydrogen chloride 1/1
53486469
(3S)-3-methylmorpholine;hydrochloride
CC1COCCN1.Cl
Melting Point 121.0°C to 124.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 137.61
Percent Purity 97%
Chemical Name or Material (S)-3-Methylmorpholine hydrochloride
compliance-icons
Product Identifier
  • (S)-3-Methylmorpholine hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'