Läs mer
(S)-3-Phenyl-Beta-alanine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-3-Phenyl-beta-alanine Hydrochloride |
| CAS | 83649-47-2 |
| Molekylformel | C9 H11 N O2 . Cl H |
| Kvantitet | 5 g |
| Synonym | Benzenepropanoic acid','β-amino-','hydrochloride (1:1)','(βS)-, Benzenepropanoic acid','β-amino-','hydrochloride','(S)-, Benzenepropanoic acid','β-amino-','hydrochloride','(βS)- (9CI), (+)-(3S)-3-Amino-3-phenylpropionic acid hydrochloride, (-)-3-Amino-3-phenylpropanoic acid hydrochloride, (S)-(-)-3-Amino-3-phenylpropionic acid hydrochloride, (S)-3-Amino-3-phenylpropanoic acid hydrochloride, (S)-3-Amino-3-phenylpropionic acid hydrochloride, (S)-3-Phenyl-β-alanine hydrochloride |
| InChI formel | InChI=1S/C9H11NO2.ClH/c10-8(6-9(11)12)7-4-2-1-3-5-7;/h1-5,8H,6,10H2,(H,11,12);1H/t8-;/m0./s1 |
| LEDER | Cl.N[C@@H](CC(=O)O)c1ccccc1 |
| IUPAC-namn | (3S)-3-amino-3-phenylpropanoic acid;hydrochloride |
| Molekylvikt (g/mol) | 201.65 |
| Formel vikt | 201.0557 |