Läs mer
(S)-4-Hydroxy Propranolol Hydrobromide, TRC
Produktkod. 30408754
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30408754 50 mg
1 options
Produktkod. 30408754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95253350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-4-Hydroxy Propranolol Hydrochloride
CAS 135201-50-2
Molekylformel C16 H21 N O3 . Cl H
Kvantitet 50 mg
Synonym 1-Naphthalenol, 4-[(2S)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (1:1) (ACI), 1-Naphthalenol, 4-[(2S)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (9CI), 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride, (S)- (ZCI), (S)-4-Hydroxypropranolol hydrochloride, 4-[(2S)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-1-naphthalenol hydrochloride (1:1), 4-Hydroxy (S)-propranolol hydrochloride
InChI formel InChI=1S/C16H21NO3.ClH/c1-11(2)17-9-12(18)10-20-16-8-7-15(19)13-5-3-4-6-14(13)16;/h3-8,11-12,17-19H,9-10H2,1-2H3;1H/t12-;/m0./s1
LEDER Cl.CC(C)NC[C@H](O)COc1ccc(O)c2ccccc12
IUPAC-namn 4-[(2S)-2-hydroxy-3-(propan-2-ylamino)propoxy]naphthalen-1-ol;hydrochloride
Molekylvikt (g/mol) 311.8
Formel vikt 311.1288