Läs mer
(S)-4-Nitrophenyl 1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate, TRC
Produktkod. 30469736
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30469736 10 g
1 options
Produktkod. 30469736 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50449010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-4-Nitrophenyl 1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate
CAS 1229227-22-8
Molekylformel C22H18N2O4
Kvantitet 10 g
Synonym (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic Acid 4-Nitrophenyl Ester, p-Nitrophenyl (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline-2-cate
LEDER OC(=O)/C=C/c1c(C=O)cccc1
Molekylvikt (g/mol) 374.39