Läs mer
S-(4-Nitrophenyl)mercapturic Acid, TRC
Produktkod. 30464675
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464675 250 mg
30484288 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30464675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN504410250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91088-55-0
Molekylformel C11 H12 N2 O5 S
Molekylvikt (g/mol) 284.29
IUPAC-namn (2R)-2-acetamido-3-(4-nitrophenyl)sulfanylpropanoic acid
LEDER CC(=O)N[C@@H](CSc1ccc(cc1)[N+](=O)[O-])C(=O)O
Specifikationer
Kemiskt namn eller material S-(4-Nitrophenyl)mercapturic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C11H12N2O5S/c1-7(14)12-10(11(15)16)6-19-9-4-2-8(3-5-9)13(17)18/h2-5,10H,6H2,1H3,(H,12,14)(H,15,16)/t10-/m0/s1
Formel vikt 284.0467