Läs mer
(S)-4-Oxo-homophenylalanine Hydrochloride, TRC
Produktkod. 30424204
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30424204 250 mg
1 options
Produktkod. 30424204 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO856845250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-4-Oxo-homophenylalanine Hydrochloride
CAS 168154-76-5
Molekylformel C10H12ClNO3
Kvantitet 250 mg
Synonym (αS)-α-Amino-γ-oxo-benzenebutanoic Acid Hydrochloride, (S)-α-Amino-γ-oxo-benzenebutanoic Acid Hydrochloride
InChI formel InChI=1S/C10H11NO3.ClH/c11-8(10(13)14)6-9(12)7-4-2-1-3-5-7;/h1-5,8H,6,11H2,(H,13,14);1H/t8-;/m0./s1
LEDER N[C@H](C(O)=O)CC(C1=CC=CC=C1)=O.Cl
IUPAC-namn (2S)-2-amino-4-oxo-4-phenylbutanoic acid;hydrochloride
Molekylvikt (g/mol) 229.66
Formel vikt 229.0506