Läs mer
(S)-(-)-5-[(2-Acetoxypropanoyl)amino]-2,4,6-triiodoisophthalic Acid, TRC
Produktkod. 30316534
Change view
Kvantitet:
1 g
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30316534 1 g -
30314783 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30316534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1871651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-(-)-5-[(2-Acetoxypropanoyl)amino]-2,4,6-triiodoisophthalic Acid
CAS 1416451-25-6
Molekylformel C13H10I3NO7
Kvantitet 1 g
Synonym 5-[[(2S)-2-(Acetyloxy)-1-oxopropyl]amino]-2,4,6-triiodo-1,3-benzendicarboxylic Acid
InChI formel InChI=1S/C13H10I3NO7/c1-3(24-4(2)18)11(19)17-10-8(15)5(12(20)21)7(14)6(9(10)16)13(22)23/h3H,1-2H3,(H,17,19)(H,20,21)(H,22,23)/t3-/m0/s1
LEDER C[C@H](OC(=O)C)C(=O)Nc1c(I)c(C(=O)O)c(I)c(C(=O)O)c1I
IUPAC-namn 5-[[(2S)-2-acetyloxypropanoyl]amino]-2,4,6-triiodobenzene-1,3-dicarboxylic acid
Molekylvikt (g/mol) 672.93
Formel vikt 672.7591