missing translation for 'onlineSavingsMsg'
Läs mer

(S)-9-[2-(Hydroxypropyl] Adenine, TRC

Produktkod. 30404644
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30404644 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404644 Supplier Toronto Research Chemicals Supplier No. TRCH952690250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (S)-9-[2-(Hydroxypropyl)] Adenine
CAS 14047-27-9
Molekylformel C8 H11 N5 O
Kvantitet 250 mg
Synonym (S)-9-(2-Hydroxypropyl)adenine, 9-[(2S)-2-Hydroxypropyl]adenine, L-(+)-9-(2-Hydroxypropyl)adenine, (αS)-6-Amino-α-methyl-9H-purine-9-ethanol, (S)-9-[2-(Hydroxypropyl] Adenine
InChI formel InChI=1S/C8H11N5O/c1-5(14)2-13-4-12-6-7(9)10-3-11-8(6)13/h3-5,14H,2H2,1H3,(H2,9,10,11)/t5-/m0/s1
LEDER C[C@H](O)Cn1cnc2c(N)ncnc12
IUPAC-namn (2S)-1-(6-aminopurin-9-yl)propan-2-ol
Molekylvikt (g/mol) 193.206
Formel vikt 193.0964
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.