Läs mer
S-Acetyl-L-glutathione, TRC
Produktkod. 30418328
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30418328 2.5 g
1 options
Produktkod. 30418328 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2250852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S-Acetylglutathione
CAS 3054-47-5
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H19 N3 O7 S
Kvantitet 2.5 g
Synonym S-Acetyl-L-glutathione, L-γ-glutamyl-S-acetyl-L-cysteinyl-Glycine, Glutathione','S-acetate (6CI,7CI), Glutathione','acetate (ester) (8CI), Glycine','N-(S-acetyl-N-L-γ-glutamyl-L-cysteinyl)-, S-Acetylglutathione
InChI formel InChI=1S/C12H19N3O7S/c1-6(16)23-5-8(11(20)14-4-10(18)19)15-9(17)3-2-7(13)12(21)22/h7-8H,2-5,13H2,1H3,(H,14,20)(H,15,17)(H,18,19)(H,21,22)/t7-,8-/m0/s1
LEDER CC(=O)SC[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)NCC(=O)O
IUPAC-namn (2S)-5-[[(2R)-3-acetylsulfanyl-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-2-amino-5-oxopentanoic acid
Molekylvikt (g/mol) 349.36
Formel vikt 349.094