missing translation for 'onlineSavingsMsg'
Läs mer

(S)-Albuterol Hydrochloride, TRC

Produktkod. 30312687
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30312687 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30312687 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA51449025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (S)-Albuterol Hydrochloride
CAS 50293-91-9
Molekylformel C13 H21 N O3 . Cl H
Kvantitet 25 mg
Synonym (S)-Albuterol Hydrochloride
InChI formel InChI=1S/C13H21NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,12,14-17H,7-8H2,1-3H3;1H/t12-;/m1./s1
LEDER Cl.CC(C)(C)NC[C@@H](O)c1ccc(O)c(CO)c1
IUPAC-namn 4-[(1S)-2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;hydrochloride
Molekylvikt (g/mol) 275.77
Formel vikt 275.1288
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.