Läs mer
(S)-Alogliptin Benzoate Salt, TRC
Produktkod. 30328066
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30328066 100 mg
1 options
Produktkod. 30328066 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA575455100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Alogliptin Benzoate Salt
CAS 1638544-64-5
Molekylformel C18 H21 N5 O2 . C7 H6 O2
Kvantitet 100 mg
Synonym 2-[[6-[(3S)-3-Amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-benzonitrile Benzoate
InChI formel InChI=1S/C18H21N5O2.C7H6O2/c1-21-17(24)9-16(22-8-4-7-15(20)12-22)23(18(21)25)11-14-6-3-2-5-13(14)10-19;8-7(9)6-4-2-1-3-5-6/h2-3,5-6,9,15H,4,7-8,11-12,20H2,1H3;1-5H,(H,8,9)/t15-;/m0./s1
LEDER CN1C(=O)C=C(N2CCC[C@H](N)C2)N(Cc3ccccc3C#N)C1=O.OC(=O)c4ccccc4
IUPAC-namn 2-[[6-[(3S)-3-aminopiperidin-1-yl]-3-methyl-2,4-dioxopyrimidin-1-yl]methyl]benzonitrile;benzoic acid
Molekylvikt (g/mol) 461.513
Formel vikt 461.206