Läs mer
(S)-Alpha-Glycerophosphocholine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-alpha-Glycerophosphocholine |
| CAS | 4217-84-9 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C8H20NO6P |
| Kvantitet | 25 mg |
| Synonym | L-Choline hydroxide 2,3-Dihydroxypropyl Hydrogen Phosphate inner Salt, (S)-2-[[(2,3-Dihydroxypropoxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-ethanaminium Inner Salt, D-Glycerolphosphocholine, Glycerophosphocholine, Glycerophosphorylcholine, alpha-GPC, [(2S)-2,3-Dihydroxypropyl] 2-trimethylazaniumylethyl Phosphate |
| InChI formel | InChI=1S/C8H20NO6P/c1-9(2,3)4-5-14-16(12,13)15-7-8(11)6-10/h8,10-11H,4-7H2,1-3H3/t8-/m0/s1 |
| LEDER | C[N+](C)(C)CCOP(=O)([O-])OC[C@@H](O)CO |
| IUPAC-namn | [(2S)-2,3-dihydroxypropyl] 2-(trimethylazaniumyl)ethyl phosphate |
| Molekylvikt (g/mol) | 257.22 |
| Visa mer |