missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
250 mg
1 g
Description
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.
Solubility
Slightly miscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.
Solubility
Slightly miscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Density | 1.353 |
Boiling Point | 213°C to 214°C |
Flash Point | 89°C (192°F) |
Refractive Index | 1.469 |
Quantity | 50 mg |
UN Number | UN3265 |
Beilstein | 3591564 |
Sensitivity | Moisture sensitive |
Merck Index | 14,6280 |
Solubility Information | Slightly miscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'