Läs mer
(S)-Alpha-Methylhippuric Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-alpha-Methylhippuric Acid |
| CAS | 2198-64-3 |
| Molekylformel | C10H11NO3 |
| Kvantitet | 5 g |
| Synonym | N-Benzoyl-L-alanine, (S)-N-Benzoylalanine, L-N-Benzoylalanine, N-Benzoyl-L-alanine, N-Benzoyl-alpha-alanine, N-Benzoylalanine, NSC 167298, NSC 43124, alpha-Methylhippuric Acid |
| InChI formel | InChI=1S/C10H11NO3/c1-7(10(13)14)11-9(12)8-5-3-2-4-6-8/h2-7H,1H3,(H,11,12)(H,13,14)/t7-/m0/s1 |
| LEDER | C[C@H](NC(=O)c1ccccc1)C(=O)O |
| IUPAC-namn | (2S)-2-benzamidopropanoic acid |
| Molekylvikt (g/mol) | 193.2 |
| Formel vikt | 193.0739 |