Läs mer
(S)-(-)-Anatabine, TRC
Produktkod. 30007373
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30007373 25 mg -
30008734 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007373 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63749525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 581-49-7
Molekylformel *C6C4H12N2
Molekylvikt (g/mol) 166.2
Synonym 2,3'-Bipyridine, 1,2,3,6-tetrahydro-, (2S)- (9CI, ACI), (2S)-1,2,3,6-Tetrahydro-2,3'-bipyridine (ACI), 2,3'-Bipyridine, 1,2,3,6-tetrahydro-, (S)- (ZCI), Anatabine (6CI, 7CI, 8CI), (-)-Anatabine, (S)-(-)-Anatabine, (S)-Anatabine, 3-[(2S)-1,2,3,6-Tetrahydropyridin-2-yl]pyridine, Anatabine, (-)-
IUPAC-namn 3-[(2S)-1,2,3,6-tetrahydropyridin-2-yl]pyridine
LEDER C1N[C@@H](CC=C1)c2cccnc2
Specifikationer
Kemiskt namn eller material (S)-(-)-Anatabine
Kvantitet 25 mg
InChI formel InChI=1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-4,6,8,10,12H,5,7H2/t10-/m0/s1
Formel vikt 160.1