Läs mer
S-(+)-Atomoxetine Hydrochloride, TRC
Produktkod. 30329829
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30329829 50 mg
1 options
Produktkod. 30329829 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79140550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material ent S-(+) Atomoxetine HCl
CAS 82857-39-4
Molekylformel C17 H21 N O . Cl H
Kvantitet 50 mg
Synonym (γS)-N-methyl-γ-(2-methylphenoxy)-Benzenepropanamine hydrochloride (1:1), Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (S)-, Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (γS)- (9CI), (S)-(+)-Tomoxetine hydrochloride, (S)-Tomoxetine hydrochloride, LY 139602, Atomoxetine Hydrochloride Imp B (EP) as HCl
InChI formel InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m0./s1
LEDER Cl.CNCC[C@H](Oc1ccccc1C)c2ccccc2
IUPAC-namn (3S)-N-methyl-3-(2-methylphenoxy)-3-phenylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 291.82
Formel vikt 291.139