Läs mer
(S)-Benzyl 2-(5-Bromo-1H-indole-3-carbonyl)pyrrolidine-1-carboxylate, TRC
Produktkod. 30330112
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30330112 100 mg
1 options
Produktkod. 30330112 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB688770100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Benzyl 2-(5-Bromo-1H-indole-3-carbonyl)pyrrolidine-1-carboxylate
CAS 199659-03-5
Molekylformel C21H19BrN2O3
Kvantitet 100 mg
Synonym 1-Pyrrolidinecarboxylic acid, 2-[(5-bromo-1H-indol-3-yl)carbonyl]-, phenylmethyl ester, (S)- (ZCI), Phenylmethyl (2S)-2-[(5-bromo-1H-indol-3-yl)carbonyl]-1-pyrrolidinecarboxylate (ACI)
InChI formel InChI=1S/C21H19BrN2O3/c22-15-8-9-18-16(11-15)17(12-23-18)20(25)19-7-4-10-24(19)21(26)27-13-14-5-2-1-3-6-14/h1-3,5-6,8-9,11-12,19,23H,4,7,10,13H2/t19-/m0/s1
LEDER O=C(N1[C@H](C(C2=CNC3=C2C=C(Br)C=C3)=O)CCC1)OCC4=CC=CC=C4
IUPAC-namn benzyl (S)-2-(5-bromo-1H-indole-3-carbonyl)pyrrolidine-1-carboxylate
Molekylvikt (g/mol) 427.29
Formel vikt 426.06