Läs mer
(S)-Benzyl 6-Methyloctanoate, TRC
Produktkod. 30344198
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30344198 1 mg
1 options
Produktkod. 30344198 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2857281MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Benzyl 6-Methyloctanoate
CAS 2381968-90-5
Molekylformel C16H24O2
Kvantitet 1 mg
Synonym Octanoic acid, 6-methyl-, phenylmethyl ester,
InChI formel InChI=1S/C16H24O2/c1-3-14(2)9-7-8-12-16(17)18-13-15-10-5-4-6-11-15/h4-6,10-11,14H,3,7-9,12-13H2,1-2H3/t14-/m0/s1
LEDER O=C(CCCC[C@@H](C)CC)OCC1=CC=CC=C1
IUPAC-namn (S)-Benzyl 6-Methyloctanoate
Molekylvikt (g/mol) 248.36
Formel vikt 248.13