Läs mer
S-Benzyl di-sec-Butylthiocarbamate (Tiocarbazil), TRC
Produktkod. 30341677
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30341677 100 mg
1 options
Produktkod. 30341677 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB705590100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tiocarbazil
CAS 36756-79-3
Molekylformel C16 H25 N O S
Kvantitet 100 mg
Synonym Carbamothioic acid, bis(1-methylpropyl)-, S-(phenylmethyl) ester (9CI), Drepamon, M 3432, S-Benzyl di-sec-butylthiocarbamate, Thiocarbazil, Tiocarbazil, Carbamothioic acid, N,N-bis(1-methylpropyl)-, S-(phenylmethyl) ester
InChI formel InChI=1S/C16H25NOS/c1-5-13(3)17(14(4)6-2)16(18)19-12-15-10-8-7-9-11-15/h7-11,13-14H,5-6,12H2,1-4H3
LEDER CCC(C)N(C(C)CC)C(=O)SCc1ccccc1
IUPAC-namn S-benzyl N,N-di(butan-2-yl)carbamothioate
Molekylvikt (g/mol) 279.44
Formel vikt 279.1657