Läs mer
S-Benzyl-L-homocysteine, TRC
Produktkod. 30342068
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30342068 500 mg
1 options
Produktkod. 30342068 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB278800500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S-Benzyl-L-homocysteine
CAS 7689-60-3
Molekylformel C11 H15 N O2 S
Kvantitet 500 mg
Synonym L-2-Amino-4-(benzylthio)butyric Acid, S-(Phenylmethyl)-L-homocysteine
InChI formel InChI=1S/C11H15NO2S/c12-10(11(13)14)6-7-15-8-9-4-2-1-3-5-9/h1-5,10H,6-8,12H2,(H,13,14)/t10-/m0/s1
LEDER N[C@@H](CCSCc1ccccc1)C(=O)O
IUPAC-namn (2S)-2-amino-4-benzylsulfanylbutanoic acid
Molekylvikt (g/mol) 225.31
Formel vikt 225.0823