missing translation for 'onlineSavingsMsg'
Läs mer

(S)-beta-Butyrolactone, TRC

Produktkod. 30346655
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30346655 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30346655 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB761680100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (S)-Beta-Butyrolactone
CAS 65058-82-4
Molekylformel C4 H6 O2
Kvantitet 100 mg
Synonym (4S)-4-Methyl-2-oxetanone, (-)-β-Butyrolactone, (-)-β-Methylpropiolactone, (4s)-4-Methyloxetan-2-one, (S)-4-Methyloxetan-2-one, (S)-Butyrolactone, (S)-4-Methyloxetan-2-one
InChI formel InChI=1S/C4H6O2/c1-3-2-4(5)6-3/h3H,2H2,1H3/t3-/m0/s1
LEDER C[C@H]1CC(=O)O1
IUPAC-namn (4S)-4-methyloxetan-2-one
Molekylvikt (g/mol) 86.089
Formel vikt 86.037
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.