Läs mer
(S)-(-)-Bupivacaine Hydrochloride, TRC
Produktkod. 30137217
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137217 500 mg
30134602 10 mg
30012413 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137217 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB689675500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27262-48-2
Molekylformel C18 H28 N2 O . Cl H
Molekylvikt (g/mol) 324.89
Synonym 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, hydrochloride (1:1), (2S)-, 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, monohydrochloride, (2S)- (9CI), 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, monohydrochloride, (S)-, 2',6'-Pipecoloxylidide, 1-butyl-, monohydrochloride, (-)- (8CI), (-)-Bupivacaine monohydrochloride, (S)-(-)-Bupivacaine monohydrochloride, Chirocaine, Levobupivacaine hydrochloride, Popscaine, (2S)-1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide hydrochloride, (S)-Bupivacaine HCl (Levobupivacaine HCl)
IUPAC-namn (2S)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride
LEDER Cl.CCCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C
Specifikationer
Kemiskt namn eller material (S)-(-)-Bupivacaine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C18H28N2O.ClH/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H/t16-;/m0./s1
Formel vikt 324.1968
Procent renhet >95%