Läs mer
S(-)-BZM Hydrochloride, TRC
Produktkod. 30347865
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30347865 10 mg
1 options
Produktkod. 30347865 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB80280110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S(-)-BZM Hydrochloride
CAS 132448-73-8
Molekylformel C15H23ClN2O3
Kvantitet 10 mg
Synonym N-[[(2S)-1-Ethyl-2-pyrrolidinyl]methyl]-2-hydroxy-6-methoxybenzamide Hydrochloride, (S)-N-[(1-Ethyl-2-pyrrolidinyl)methyl]-2-hydroxy-6-methoxybenzamide Hydrochloride, (S)-N-[(1-Ethyl-2-pyrrolidinyl)methyl]-2-hydroxy-6-methoxybenzamide Monohydrochloride
InChI formel InChI=1S/C15H22N2O3.ClH/c1-3-17-9-5-6-11(17)10-16-15(19)14-12(18)7-4-8-13(14)20-2;/h4,7-8,11,18H,3,5-6,9-10H2,1-2H3,(H,16,19);1H/t11-;/m0./s1
LEDER Cl.CCN1CCC[C@H]1CNC(=O)c2c(O)cccc2OC
IUPAC-namn N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide;hydrochloride
Molekylvikt (g/mol) 314.81
Formel vikt 314.1397