Läs mer
(S)-(-)-Canadine, TRC
Produktkod. 17962055
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962055 250 mg
17953205 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17962055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC175180250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5096-57-1
Molekylformel C20 H21 N O4
Molekylvikt (g/mol) 339.38
Synonym (S)-5,8,13,13a-tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, Canadine, (-)-Canadine, (-)-Tetrahydroberberine, (-)-α-Canadine, (S)-(-)-7,8,13,13a-Tetrahydroberberine, (S)-(-)-Tetrahydroberberine, (S)-Canadine, (S)-Tetrahydroberberine, 9,10-Dimethoxy-2,3-(methylenedioxy)-13aα-berbine, l-Canadine, α-Canadine
IUPAC-namn (1S)-16,17-dimethoxy-5,7-dioxa-13-azapentacyclo[11.8.0.02,10.04,8.015,20]henicosa-2,4(8),9,15(20),16,18-hexaene
LEDER COc1ccc2C[C@@H]3N(CCc4cc5OCOc5cc34)Cc2c1OC
Specifikationer
Kemiskt namn eller material (S)-(-)-Canadine
Kvantitet 250 mg
InChI formel InChI=1S/C20H21NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,8-9,16H,5-7,10-11H2,1-2H3/t16-/m0/s1
Formel vikt 339.1471