Läs mer
(S)-Carbidopa Methyl Ester (>90%), TRC
Produktkod. 30366852
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366852 100 mg
30350529 50 mg
30358156 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366852 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC176025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52514-63-3
Molekylformel C11 H16 N2 O4
Molekylvikt (g/mol) 240.256
Synonym (S)-α-Hydrazino-3,4-dihydroxy-α-methyl-benzenepropanoic acid methyl ester, (S)-alpha-Hydrazino-3,4-dihydroxy-alpha-methylbenzenepropanoic Acid Methyl Ester
IUPAC-namn methyl (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoate
LEDER COC(=O)[C@](C)(Cc1ccc(O)c(O)c1)NN
Specifikationer
Kemiskt namn eller material Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Methyl Ester)
Kvantitet 100 mg
InChI formel InChI=1S/C11H16N2O4/c1-11(13-12,10(16)17-2)6-7-3-4-8(14)9(15)5-7/h3-5,13-15H,6,12H2,1-2H3/t11-/m0/s1
Formel vikt 240.111