Läs mer
(S)-Carbidopa Methyl Ester N-Cyclohexylidene, TRC
Produktkod. 30364684
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364684 10 mg
30359605 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364684 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17601510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 934371-48-9
Molekylformel C17 H24 N2 O4
Molekylvikt (g/mol) 320.38
Synonym Methyl (2S)-2-(2-Cyclohexylidenehydrazino)-3-(3,4-dihydroxyphenyl)-2-methyl-propanoate
IUPAC-namn methyl (2S)-2-(2-cyclohexylidenehydrazinyl)-3-(3,4-dihydroxyphenyl)-2-methylpropanoate
LEDER COC(=O)[C@](C)(Cc1ccc(O)c(O)c1)NN=C2CCCCC2
Specifikationer
Kemiskt namn eller material (S)-Carbidopa methyl ester N-Cyclohexylidene
Kvantitet 10 mg
InChI formel InChI=1S/C17H24N2O4/c1-17(16(22)23-2,19-18-13-6-4-3-5-7-13)11-12-8-9-14(20)15(21)10-12/h8-10,19-21H,3-7,11H2,1-2H3/t17-/m0/s1
Formel vikt 320.1736