missing translation for 'onlineSavingsMsg'
Läs mer

(S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide, Mikromol™

Produktkod. 17989114
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17989114 25 mg
17347902 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17989114 Leverantör Mikromol Leverantörsnummer MM0236.010025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
CAS 5439-87-2
Analyt- eller komponentnamn (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
Frakt skick Room Temperature
Molekylformel C5 H9 N O5 S
Certifikat och efterlevnad ISO 17025
Synonym (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
InChI formel InChI=1S/C5H9NO5S/c6-3(5(9)10)1-12(11)2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-,12?/m0/s1
LEDER N[C@@H](CS(=O)CC(=O)O)C(=O)O
IUPAC-namn (2R)-2-amino-3-(carboxymethylsulfinyl)propanoic acid
Molekylvikt (g/mol) 195.19
Kvantitet 25 mg
Formel vikt 195.0201 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.