Läs mer
(S)-Carnitine Mesylate Isobutylester, Mesylate Salt, TRC
Produktkod. 30351395
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30351395 50 mg
1 options
Produktkod. 30351395 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18412550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Carnitine Mesylate Isobutylester, Mesylate Salt
CAS 161886-59-5
Molekylformel C12 H26 N O5 S . C H3 O3 S
Kvantitet 50 mg
Synonym 1-Butanaminium, N,N,N-trimethyl-4-(2-methylpropoxy)-2-[(methylsulfonyl)oxy]-4-oxo-, (S)-, methanesulfonate (9CI), (S)-Carnitine Mesylate Isobutylester, Mesylate Salt
InChI formel InChI=1S/C12H26NO5S.CH4O3S/c1-10(2)9-17-12(14)7-11(8-13(3,4)5)18-19(6,15)16;1-5(2,3)4/h10-11H,7-9H2,1-6H3;1H3,(H,2,3,4)/q+1;/p-1/t11-;/m0./s1
LEDER CC(C)COC(=O)C[C@@H](C[N+](C)(C)C)OS(=O)(=O)C.CS(=O)(=O)[O-]
IUPAC-namn methanesulfonate;trimethyl-[(2S)-4-(2-methylpropoxy)-2-methylsulfonyloxy-4-oxobutyl]azanium
Molekylvikt (g/mol) 391.5
Formel vikt 391.1335