Läs mer
S-(-)-Cotinine Perchlorate, TRC
Produktkod. 30361980
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30361980 250 mg
1 options
Produktkod. 30361980 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC725015250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S-(-)-Cotinine Perchlorate
CAS 1217692-15-3
Molekylformel C10 H12 N2 O . Cl H O4
Kvantitet 250 mg
Synonym (-)-Cotinine Perchlorate, S-(-)-Cotinine Perchlorate
InChI formel InChI=1S/C10H12N2O.ClHO4/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8;2-1(3,4)5/h2-3,6-7,9H,4-5H2,1H3;(H,2,3,4,5)/t9-;/m0./s1
LEDER CN1[C@@H](CCC1=O)c2cccnc2.OCl(=O)(=O)=O
IUPAC-namn (5S)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one;perchloric acid
Molekylvikt (g/mol) 276.67
Formel vikt 276.0513