Learn More
Cole-Parmer™ Stuart™ Polypropylene Bottle Top Dispenser
Designed for routine dispensing of common laboratory liquids such as dilute acids, buffers, biological solutions, polar solvents and strong alkaline solutions. Stuart™ Polypropylene Bottle Top Dispenser provides a low cost option for liquid dispensing without compromising accuracy or reliability.
Brand:  Cole-Parmer™ BD25
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12073893
  • 3123.57 SEK / Each
Estimated Shipment: 07-10-2021
Add to basket
Description
Description
  • Economical dispensing of non-aggressive liquids
  • Low-cost replaceable piston/cylinder and valves
  • Easy cleaning and maintenance
  • Simple to operate
  • Automatic refill
Specifications
Specifications
1.2%
Dispenser
Polypropylene
BD25
0.2%
2.5 to 25 mL
Closing aqueous solutions, alkalis, dilute acids and polar solvents such as ethanol or acetone
0.5 mL
Videos
SDS
Documents
Certificates
View accessory products