Läs mer
(S)-Desmethyl Citalopram Ethanedioate, TRC
Produktkod. 30358101
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30358101 50 mg
1 options
Produktkod. 30358101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29151550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trityl Olmesartan
CAS 852172-06-6
Molekylformel C19 H19 F N2 O . C2 H2 O4
Kvantitet 50 mg
Synonym 5-Isobenzofurancarbonitrile, 1-(4-fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-, (1S)-, ethanedioate (1:1) (9CI), (1S)-1-(4-Fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-5-isobenzofurancarbonitrile oxalate, (S)-1-(3-Methylaminopropyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile oxalate, (S)-Desmethylcitalopram oxalate
InChI formel InChI=1S/C19H19FN2O.C2H2O4/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;3-1(4)2(5)6/h3-8,11,22H,2,9-10,13H2,1H3;(H,3,4)(H,5,6)/t19-;/m0./s1
LEDER CNCCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O
IUPAC-namn (1S)-1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3H-2-benzofuran-5-carbonitrile;oxalic acid
Molekylvikt (g/mol) 400.4
Formel vikt 400.1434