Läs mer
S-Desmethyl-S-(2-hydroxy-2-methylpropyl) Rosuvastatin Sodium, TRC
Produktkod. 30132821
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132821 100 mg
30011378 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132821 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD291735100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1714147-50-8
Molekylformel C25 H33 F N3 O7 S . Na
Molekylvikt (g/mol) 561.6
Synonym (3R,5S,6E)-7-[4-(4-Fluorophenyl)-2-[[(2-hydroxy-2-methylpropyl)sulfonyl]methylamino]-6-(1-methylethyl)-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid Sodium Salt (1:1), Rosuvastatin Calcium Imp. A (EP) as Sodium Salt, (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl[(2-hydroxy-2-methylpropyl)sulfonyl]amino]-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid Sodium, Rosuvastatin EP Impurity A;
IUPAC-namn sodium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[(2-hydroxy-2-methylpropyl)sulfonyl-methylamino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
LEDER [Na+].CC(C)c1nc(nc(c2ccc(F)cc2)c1\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])N(C)S(=O)(=O)CC(C)(C)O
Specifikationer
Kemiskt namn eller material (3R,5S,6E)-7-(4-(4-Fluorophenyl)-2-(((2-hydroxy-2-methylpropyl)sulfonyl)(methyl)amino)-6-(1-methylethyl)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoic Acid Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C25H34FN3O7S.Na/c1-15(2)22-20(11-10-18(30)12-19(31)13-21(32)33)23(16-6-8-17(26)9-7-16)28-24(27-22)29(5)37(35,36)14-25(3,4)34;/h6-11,15,18-19,30-31,34H,12-14H2,1-5H3,(H,32,33);/q;+1/p-1/b11-10+;/t18-,19-;/m1./s1
Formel vikt 561.1921
Procent renhet >95%