Läs mer
S-Desmethyl-S-(2-hydroxy-2-methylpropyl) Rosuvastatin tert-Butyl Ester, TRC
Produktkod. 30365917
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365917 25 mg
30349527 10 mg
30360564 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29175025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1714147-49-5
Molekylformel C29H42FN3O7S
Molekylvikt (g/mol) 595.72
Synonym Rosuvastatin EP Impurity A T-Butyl Ester
LEDER FC1=CC=C(C2=NC(N(C)S(=O)(CC(O)(C)C)=O)=NC(C(C)C)=C2/C=C\C(O)CC(O)CC(OC(C)(C)C)=O)C=C1
Specifikationer
Kemiskt namn eller material S-Desmethyl-S-(2-hydroxy-2-methylpropyl) Rosuvastatin tert-Butyl Ester
Kvantitet 25 mg
InChI formel InChI=1S/C29H42FN3O7S/c1-18(2)25-23(14-13-21(34)15-22(35)16-24(36)40-28(3,4)5)26(19-9-11-20(30)12-10-19)32-27(31-25)33(8)41(38,39)17-29(6,7)37/h9-14,18,21-22,34-35,37H,15-17H2,1-8H3/b14-13+
Formel vikt 595.2727