Läs mer
(S)-(-)-Dropropizine N,N-Dioxide, TRC
Produktkod. 30385710
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30385710 100 mg
1 options
Produktkod. 30385710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD681535100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-(-)-Dropropizine N,N-Dioxide
CAS 2098198-51-5
Molekylformel C13 H20 N2 O4
Kvantitet 100 mg
Synonym (2S)-3-(1,4-dioxido-4-phenyl-1-piperazinyl)-1,2-propanediol
InChI formel InChI=1S/C13H20N2O4/c16-11-13(17)10-14(18)6-8-15(19,9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-,14?,15?/m0/s1
LEDER OC[C@@H](O)CN1(=O)CCN(=O)(CC1)c2ccccc2
IUPAC-namn (2S)-3-(1,4-dioxo-4-phenyl-1$l^{5},4$l^{5}-diazinan-1-yl)propane-1,2-diol
Molekylvikt (g/mol) 268.31
Formel vikt 268.1423