Läs mer
(S)-(-)-Dropropizine N-Oxide, TRC
Produktkod. 30133739
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133739 250 mg
17923675 25 mg
17922565 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133739 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD681530250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 152237-40-6
Molekylformel C13 H20 N2 O3
Molekylvikt (g/mol) 252.31
Synonym 1,2-Propanediol, 3-(1-oxido-4-phenyl-1-piperazinyl)-, (S)- (9CI), 1,2-Propanediol, 3-(4-phenyl-1-piperazinyl)-, N-oxide, (S)-, Levodropropizine N-Oxide
IUPAC-namn (2S)-3-(1-oxido-4-phenylpiperazin-1-ium-1-yl)propane-1,2-diol
LEDER OC[C@@H](O)C[N+]1([O-])CCN(CC1)c2ccccc2
Specifikationer
Kemiskt namn eller material Levodropropizine N-Oxide
Kvantitet 250 mg
InChI formel InChI=1S/C13H20N2O3/c16-11-13(17)10-15(18)8-6-14(7-9-15)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m0/s1
Formel vikt 252.1474