missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
(S)-Duloxetine Phthalamide, Mikromol
Beskrivning
(S)-Duloxetine Phthalamide, Mikromol
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-Duloxetine Phthalamide |
| CAS | 199191-67-8 |
| Analyt- eller komponentnamn | (S)-Duloxetine Phthalamide |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C26 H23 N O4 S |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Benzoic acid, 2-[[methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]carbonyl]-, 2-[[Methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]carbonyl]benzoic acid |
| InChI formel | InChI=1S/C26H23NO4S/c1-27(25(28)20-11-4-5-12-21(20)26(29)30)16-15-23(24-14-7-17-32-24)31-22-13-6-9-18-8-2-3-10-19(18)22/h2-14,17,23H,15-16H2,1H3,(H,29,30)/t23-/m0/s1 |
| Lösningstyp | Neat |
| LEDER | CN(CC[C@H](Oc1cccc2ccccc12)c3cccs3)C(=O)c4ccccc4C(=O)O |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?