Läs mer
(S)-Duloxetine Phthalamide, TRC
Produktkod. 17980965
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17980965 100 mg
17933055 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17980965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD721020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 199191-67-8
Molekylformel C26 H23 N O4 S
Molekylvikt (g/mol) 445.53
Synonym Benzoic acid, 2-[[methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]carbonyl]-, 2-[[Methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]carbonyl]benzoic acid, (S)-Duloxetine Phthalamide, Benzoic acid, 2-[[methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]carbonyl]-, 2-[[Methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]carbonyl]benzoic acid
IUPAC-namn 2-[methyl-[(3S)-3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl]carbamoyl]benzoic acid
LEDER CN(CC[C@H](Oc1cccc2ccccc12)c3cccs3)C(=O)c4ccccc4C(=O)O
Specifikationer
Kemiskt namn eller material (S)-Duloxetine Phthalamide
Kvantitet 100 mg
InChI formel InChI=1S/C26H23NO4S/c1-27(25(28)20-11-4-5-12-21(20)26(29)30)16-15-23(24-14-7-17-32-24)31-22-13-6-9-18-8-2-3-10-19(18)22/h2-14,17,23H,15-16H2,1H3,(H,29,30)/t23-/m0/s1
Formel vikt 445.1348