missing translation for 'onlineSavingsMsg'
Läs mer

(S)-(-)-Ethyl Lactate, Sigma-Aldrich™

Produktkod. 15876700
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mL
1 L
Förpackningsstorlek:
1 L
100ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15876700 1 L 1 L
15871661 100 mL 100ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15876700 Leverantör Sigma Aldrich Leverantörsnummer 8221001000
 Begränsad artikel
Beräknad leverans: 25-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-47-8
Kvalitet Research
Molekylformel CH3CH(OH)COOC2H5
Molekylvikt (g/mol) 118.13

Specifications

Smältpunkt -25°C
Densitet 1.03 g/cm3 at 20°C
Kokpunkt 154°C/1013 hPa
MDL-nummer MFCD00004518
Kvantitet 1 L
Synonym (S)-(-)-Ethyl lactate, L(-)-Lactic acid ethyl ester, (S)-(-)-2-Hydroxypropionic acid ethyl ester, (S)-(-)-Ethyl lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.