Läs mer
(S)-Flurbiprofen (S)-(-)-Alpha-Methylbenzylamine Salt, TRC
Produktkod. 30407395
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30407395 1 g
1 options
Produktkod. 30407395 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5987161G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Flurbiprofen (S)-(-)-alpha-Methylbenzylamine Salt
CAS 76319-57-8
Molekylformel C15H13FO2 C8H11N
Kvantitet 1 g
Synonym (2S)-2-(2-Fluoro-1,1'-biphenyl-4-yl)propanoic Acid (S)-(-)-alpha-Methylbenzylamine Salt, (2S)-2-(2-Fluoro-4-biphenyl)propanoic Acid (S)-(-)-alpha-Methylbenzylamine Salt
InChI formel InChI=1S/C15H13FO2.C8H11N/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11;1-7(9)8-5-3-2-4-6-8/h2-10H,1H3,(H,17,18);2-7H,9H2,1H3/t10-;7-/m00/s1
LEDER C[C@H](N)c1ccccc1.C[C@H](C(=O)O)c2ccc(c(F)c2)c3ccccc3
IUPAC-namn (2S)-2-(3-fluoro-4-phenylphenyl)propanoic acid;(1S)-1-phenylethanamine
Molekylvikt (g/mol) 365.4
Formel vikt 365.1791