Läs mer
(S)-Gossypol (acetic acid), MedChemExpress
Produktkod. 16462637
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16462637 50 mg 50mg
16436548 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16462637 Leverantör Medchem Express Leverantörsnummer HY15464D
MedChemExpress (S)-Gossypol is the isomer of a natural product Gossypol. (S)-Gossypol binds to the BH3-binding groove of Bcl-xL and Bcl-2 proteins with high affinity.
(S)-Gossypol is the isomer of a natural product Gossypol. (S)-Gossypol binds to the BH3-binding groove of Bcl-xL and Bcl-2 proteins with high affinity.
Specifikationer
Kemiskt namn eller material (S)-Gossypol (acetic acid)
Fysisk form Solid
CAS 1189561-66-7
Färg Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C32H34O10
För användning med (applikation) Cancer-programmed cell death
Synonym (S)-(+)-Gossypol acetic acid
Löslighetsinformation DMSO : 50 mg/mL (86.41 mM; Need ultrasonic)
LEDER OC1=C2C(C=O)=C(O)C(O)=C(C(C)C)C2=CC(C)=[C@@]1[C@@]3=C(C)C=C4C(C(C)C)=C(O)C(O)=C(C=O)C4=C3O.CC(O)=O.[S]
Molekylvikt (g/mol) 578.61
Formel vikt 578.61
Procent renhet 99.01%