Läs mer
(S)-(+)-Ibuprofen (S)-(+)-Lysinate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-(+)-Ibuprofen (S)-(+)-Lysinate |
| CAS | 113403-10-4 |
| Molekylformel | C13 H18 O2 . C6 H14 N2 O2 |
| Kvantitet | 250 mg |
| Synonym | L-Lysine, (αS)-α-methyl-4-(2-methylpropyl)benzeneacetate (1:1) (ACI), L-Lysine, mono[(S)-α-methyl-4-(2-methylpropyl)benzeneacetate] (ZCI), L-Lysine, mono[(αS)-α-methyl-4-(2-methylpropyl)benzeneacetate] (9CI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (S)-, compd. with L-lysine (1:1) (ZCI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (αS)-, compd. with L-lysine (1:1) (9CI), (S)-Ibuprofen-(S)-lysine, Dexibuprofen lysinate, Dexibuprofen lysine, L 669445, L 669455, MK 233, S-(+)-Ibuprofen L-lysine salt, S-Ibuprofen lysinate |
| InChI formel | InChI=1S/C13H18O2.C6H14N2O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;7-4-2-1-3-5(8)6(9)10/h4-7,9-10H,8H2,1-3H3,(H,14,15);5H,1-4,7-8H2,(H,9,10)/t10-;5-/m00/s1 |
| LEDER | CC(C)Cc1ccc(cc1)[C@H](C)C(=O)O.NCCCC[C@H](N)C(=O)O |
| IUPAC-namn | (2S)-2,6-diaminohexanoic acid;(2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid |
| Molekylvikt (g/mol) | 352.47 |
| Formel vikt | 352.2362 |