missing translation for 'onlineSavingsMsg'
Läs mer

(S)-Isopropyl-2-aminopropanoate Hydrochloride, TRC

Produktkod. 30404995
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30404995 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30404995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI822220250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (S)-Isopropyl-2-aminopropanoate Hydrochloride
CAS 62062-65-1
Molekylformel C6 H13 N O2 . H Cl
Kvantitet 250 mg
Synonym L-Alanine Isopropyl Ester Hydrochloride, Isopropyl L-Alaninate Hydrochloride, L-Alanine 2-Propyl Ester Hydrochloride, L-Alanine Isopropyl Ester Hydrochloride, O-Isopropyl-L-alanine Hydrochloride
InChI formel InChI=1S/C6H13NO2.ClH/c1-4(2)9-6(8)5(3)7;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1
LEDER Cl.CC(C)OC(=O)[C@H](C)N
IUPAC-namn propan-2-yl (2S)-2-aminopropanoate;hydrochloride
Molekylvikt (g/mol) 167.634
Formel vikt 167.071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.