Läs mer
(S)-Ketorolac, TRC
Produktkod. 30394311
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30394311 10 mg
30403498 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30394311 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK23560510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66635-92-5
Molekylformel C15H13NO3
Molekylvikt (g/mol) 255.27
Synonym 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, (1S)-, 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, (S)-, (1S)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid, (-)-Ketorolac, (S)-Ketorolac, S-(-)-Ketorolac
LEDER OC(=O)[C@H]1CCn2c1ccc2C(=O)c3ccccc3
Specifikationer
Kemiskt namn eller material (S)-Ketorolac
Kvantitet 10 mg
InChI formel InChI=1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13/h1-7,11H,8-9H2,(H,18,19)/t11-/m0/s1
Formel vikt 255.0895