Läs mer
(S)-Lisinopril Dimer, TRC
Produktkod. 30465057
Change view
Kvantitet:
0.25 mg
Produktkod. Kvantitet
30465057 0.25 mg
1 options
Produktkod. 30465057 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4690100.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lisinopril Dimer
CAS 1356839-89-8
Molekylformel C42 H60 N6 O9
Kvantitet 0.25 mg
Synonym Lisinopril Dimer, (S)-Lisinopril Dimer, (2S)-1-[(2S)-6-Amino-2-[[(2S)-1-[[[(5S)-5-[[(1S)-1-carboxy-3-phenylpropyl]amino]-6-[(2S)-2-carboxypyrrolidin-1-yl]-6-oxohexyl]amino]carbonyl]-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic Acid, Lisinopril Dihydrate Imp. G (EP)
InChI formel InChI=1S/C42H60N6O9/c43-25-9-7-17-32(38(50)47-27-11-19-35(47)41(54)55)45-31(23-21-29-13-3-1-4-14-29)37(49)44-26-10-8-18-33(39(51)48-28-12-20-36(48)42(56)57)46-34(40(52)53)24-22-30-15-5-2-6-16-30/h1-6,13-16,31-36,45-46H,7-12,17-28,43H2,(H,44,49)(H,52,53)(H,54,55)(H,56,57)/t31-,32-,33-,34-,35-,36-/m0/s1
LEDER NCCCC[C@H](N[C@@H](CCc1ccccc1)C(=O)NCCCC[C@H](N[C@@H](CCc2ccccc2)C(=O)O)C(=O)N3CCC[C@H]3C(=O)O)C(=O)N4CCC[C@H]4C(=O)O
IUPAC-namn (2S)-1-[(2S)-6-amino-2-[[(2S)-1-[[(5S)-5-[[(1S)-1-carboxy-3-phenylpropyl]amino]-6-[(2S)-2-carboxypyrrolidin-1-yl]-6-oxohexyl]amino]-1-oxo-4-phenylbutan-2-yl]amino]hexanoyl]pyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 792.96
Formel vikt 792.4422