Läs mer
(S)-Lofexidine, TRC
Produktkod. 30474191
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30474191 2.5 mg
1 options
Produktkod. 30474191 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4694002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Lofexidine
CAS 81447-79-2
Molekylformel C11 H12 Cl2 N2 O
Kvantitet 2.5 mg
Synonym 1H-Imidazole, 2-[(1S)-1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, 1H-Imidazole, 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, (S)-, 2-[(1S)-1-(2,6-Dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole, (+)-Lofexidine, (S)-(+)-Lofexidine, Dexlofexidine
InChI formel InChI=1S/C11H12Cl2N2O/c1-7(11-14-5-6-15-11)16-10-8(12)3-2-4-9(10)13/h2-4,7H,5-6H2,1H3,(H,14,15)/t7-/m0/s1
LEDER C[C@H](Oc1c(Cl)cccc1Cl)C2=NCCN2
IUPAC-namn 2-[(1S)-1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole
Molekylvikt (g/mol) 259.13
Formel vikt 258.0327