Läs mer
(S)-(+)-Mepivacaine Hydrochloride, TRC
Produktkod. 30473148
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473148 250 mg
30470446 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30473148 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225075250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34333-71-6
Molekylformel C15 H22 N2 O . H Cl
Molekylvikt (g/mol) 282.809
Synonym (2S)-N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride, (+)-Mepivacaine Hydrochloride, (+)-1-Methyl-2',6'-pipecoloxylidide, MonoHydrochloride, (+)-Mepivacaine Hydrochloride, (S)-N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride, Ripovacaine EP Impurity C
IUPAC-namn (2S)-N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
LEDER Cl.CN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C
Specifikationer
Kemiskt namn eller material (S)-(+)-Mepivacaine Hydrochloride
Kvantitet 250 mg
InChI formel InChI=1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H/t13-;/m0./s1
Formel vikt 282.15